Perform check teaching in the User level.
For the PAL terminal button functions, refer to the PAL terminal section.
Steps
  1. On the PAL terminal screen, select Thermal Mixer 1. Select Slot 1. Select Heat. Select DWP96 ABGene.
  2. Select thermal mixer 1
    Select thermal mixer 1
    Select slot 1
    Select slot 1
    Select HEAT
    Select HEAT
  3. Select Options and select Check Teaching.
  4. Select check teaching
    Select check teaching
  5. The on-screen graphic displays the teaching points for the thermal mixer (There are three teach points: back left, front left, and front right edge positions of the module). For checking the first teach point, press Check. The robot arm moves to the teach point and checks teaching by lowering the tool to the back left edge position of the thermal mixer and verifying the correct position and coordinates along the X, Y, and Z axes. When positioned correctly, the tool aligns with the center of the teach point position.
  6. Check first teach point
    Check first teach point
    Thermal mixer first teach point
    Thermal mixer first teach point
  7. When done, press Next.
  8. Press next
    Press next
  9. Press Check to check the teaching on the second teach point at the front left edge of the thermal mixer.
  10. Check second teach point
    Check second teach point
    Thermal mixer second teach point
    Thermal mixer second teach point
    Press next
    Press next
  11. Repeat this for the third teach point at the front right. When the teaching is checked, press Next on the PAL terminal.
  12. Check third teach point
    Check third teach point
    Thermal mixer third teach point
    Thermal mixer third teach point
    Press next
    Press next
  13. Press OK. This moves the robot arm back to the home position.
Check teaching complete
Check teaching complete
NOTE
If the check teaching fails (for example, the position of the module is incorrectly aligned with respect to the tool), perform a re-teaching of the module (see Thermal mixer teaching).