Perform teaching in the Extended User level.
For the PAL terminal button functions, refer to the PAL terminal section.
Steps
- On the PAL terminal screen, select Tray Cooler 1. Select Slot 3. Select Tray A. Select DWP96 ABGene.
- Select Options and select Teach Module.
- Move the robot arm manually to the first teach point position displayed on the PAL terminal screen (back left edge of the module). Lower the tool so it touches the teach point and align it so that the tool is positioned at the center of the teach point.
- Press Save on the PAL terminal. This displays the coordinates of the saved position and allows you to manually adjust the coordinate values, if necessary. To change the coordinate values for either the x, y, or z position, select the respective co-ordinate and edit the value using the scroll wheel. When done, press Next.
- Repeat the teaching procedure for the two other teach points located at the front left edge and front right edge of the module respectively.
- When done, press Next. Press OK.