Within the content area of the PAL terminal, icons for the different tools, modules (active and passive) and I/O signals are displayed. The table below describes the available symbols.
Start Screen Icon | PALmodule Description |
|---|---|
PAL tool and Tool Change Station | |
LS Tool, Syringe Tool for liquid injections | |
HS or Headspace Tool (only PAL RSI and PAL RTC) | |
SPME Fiber Tool (only PAL RSI and PAL RTC) | |
SPME Arrow Tool (only PAL RSI and PAL RTC) | |
Dilutor Tool or Tool for Dilutor Module (only PAL RSI and PAL RTC) | |
LCMS-P Tool or Tool for LCMS-P Module (only PAL RSI and PAL RTC) | |
ITEX Tool (only PAL RSI and PAL RTC) | |
COC Tool (only PAL RSI and PAL RTC) | |
Pipette Tool (only PAL RTC) | |
Gripper Tool (only PAL RSI) | |
Park Station: Active PALmodule for automated PALtool change (only PAL RTC) | |
Active PAL module | |
Agitator | |
Vortex Mixer | |
Heatex Stirrer Module | |
Barcode Reader | |
Peltier Stack 2DW | |
Peltier Stack 6DW | |
Stack 2DW | |
Stack 6DW | |
Stack 12MT | |
Valve Drive | |
Fast Wash Module | |
Extension Module | |
SPME Fiber Conditioning Module/SPME Arrow Conditioning Module (only PAL RSI and PAL RTC) | |
OC Injector or On-column Injector (only PAL RSI and PAL RTC) | |
LCMS Wash Module (only PAL RSI and PAL RTC) | |
Dilutor Syringe Module (only PAL RSI and PAL RTC) | |
DeCapper Module | |
Centrifuge Module 2 mL | |
Centrifuge Module Combi | |
Passive PAL module | |
Tray Holder (rectangular) or Tray Plate | |
Liquid Cooled Tray Holder (Traycooler) | |
Standard Wash Module (5x10 mL Vials) | |
Large Wash Module (2 x 100 mL bottles and 1 Waste Position) | |
Solvent Module (Solvent reservoir Module for 3 x 100 mL bottles) | |
Rack: rectangular (Rack to hold vials with volume from 1 to 40 mL) | |
Vials: set of vials | |
Vial: single vial (Vial Type CV) | |
Solvent Reservoir Bottle (Vial Type SR) | |
Injector (GC Injector) | |
MHE Module (only PAL RSI and PAL RTC) | |
Tool Adapter Station | |
Synchronization and Signals | |
Input and Output Signals | |
Input Signal (GC Ready) | |
Output Signal (Injected) | |
Chromatographic System (GC) or (LC) Specified synchronization of Input and Output Signals, Cryo Trap, and Delay Time for Fake Injection for an analytical system (GC or LC). | |
General Icons | |
Logo for PAL System | |
PALrobot or PALsystem (RobotArmLeft) | |
PALhead (Injection Unit) (only PAL LSI and PAL RSI) | |