You can edit the text for item names on the PAL terminal one character at a time. The selected character is always highlighted in turquoise. Each text string is limited to a length of 15 characters and has to start with a letter.
Procedure
- Use the scroll wheel to scroll through the characters of an item name.
- The selected character is highlighted in turquoise.
Select a character - Press Enter to confirm your selection.
- On the right side of the screen, an alphabetical list of characters is displayed.
Edit a character - Use the scroll wheel to select the desired character. Press the right menu button to toggle between capital letters, small letters, or numbers and special characters.
- Press Enter to confirm your selection.
- The selected character in the name is replaced.
- Proceed with the next character that needs to be changed.
Select next character - From the Options menu, single characters can be inserted or deleted and it is possible to delete the whole text string at once.
Options menu for text strings edits