Based on the function being performed, the PAL terminal displays different menu screens with their unique options.
All menu screens have the same basic format where the screen header is the status bar, displaying the name of the current module, the access level, if a USB device is connected to the instrument, the status of the instrument (displayed by a colored LED) and the current time.
The content area shows the list of items and menus available on that particular screen. The functions associated with the menu buttons are displayed in the menu bar.
PAL terminal menu screen structure
PAL terminal menu screen structure