If the positions of any modules are found to be incorrect while checking teaching, reteach the modules using the teaching procedures described in this section.
NOTE
Full teaching is necessary only if the check teaching fails.
If you observe any errors while checking vial positions or checking teaching of any module, first verify if the vials and trays are installed correctly on the instrument interior deck before performing a teaching procedure.