At the back of the device, on the bottom right, is an ethernet connection (RJ45) and the socket for the power cord (100/240 VDC).
Power and ethernet connections on the back side of the instrument
12
No.DescriptionNo.Description
1
LAN cord for ethernet connection
2
Power cord
Steps
  1. Connect the power cord to the power socket on the back side of the instrument. The power cord specifications are as follows:
  • US cable: NEMA 5-15P to IEC 60320 C13 cable, 1.8 m, minimum rating 10A/125V
  • EU cable: Type F (CEE 7/7) to IEC 60320 C13 cable, 2.5 m, rated for minimum 10A/250V
  • A replacement cord must be rated to 10A/250 V.
  • CAUTION
    Trip hazard
    Do not use an extension cord for the instrument.
  • Connect the power cord
    Connect the power cord
  • Connect the other end of the power cord to a wall outlet.
  • (Optional) Connect the ethernet cable.
  • The ethernet cable connects the instrument to your network.