Follow the instructions in this section for proper and safe transportation and storage of the AccelerOme instrument.
WARNING
Fragile equipment
Handle with care.
The Accelerome instrument contains high-precision hardware and shock-sensitive electronic boards. Any hard impact can damage the hardware and destroy the boards.
Ensure that all personnel transporting the instrument or working with the instrument handle it carefully to avoid any kind of damage.
CAUTION
Heavy Object
The instrument weights 85 kg (187.4 pounds). Lifting it can cause muscular strain and back injury or foot injury if the instrument falls.
  • Lift the instrument with a suitable lifting device.
  • Lift the instrument with at least 4 persons.
Store the original packaging including the transport-securing foam parts for repackaging in case the instrument must be transported to a new location or sent back.