Following are the general specifications for the AccelerOme system:
Specification | Value |
|---|---|
Sample Capacity | Upto 36 label-free samples and 33 samples for isobaric mass-tagging |
Sample Capacity Temperature-controlled | Upto 36 label-free samples and 33 samples for isobaric mass-tagging |
Liquid Injection Amount | 50 uL per input sample |