The technical data of the tray cooler is as follows:
Specification | Technical data |
|---|---|
Part number | Part number of module: C0960-01-00111 |
Temperature control | 4 °C (39.2 °F) to 40 °C (104 °F) in 0.1 °C increments. Peltier element for cooling functionality of the device. For the AccelerOme system, the supported temperature range for the tray cooler is 6 °C (42.8 °F) to 17 °C (62.6 °F). |
Temperature cooling | T∆ = ≥ 14.0 °C within 60 minutes. T∆ = TAmbient temperature – TTray Cooler T∆max = 22°C (for example, for an ambient temperature of 32°C (89.6 °F), a minimum temperature of 10 °C (50 °F) can be reached). The value of TTray Cooler can be checked in the extended user level using the parameter Tray Cooler Temp. |