The technical data for the thermal mixer module is as follows:
- Orbital shaking on a circle with 1.5 mm radius from 0 to 2000 rpm
- Heating from room temperature and up to 99°C (210.2 °F)
- Accuracy:
- at 50 °C (122 °F): +/- 1.5 °C within the well plate
- at 95 °C (203 °F): +/- 1.5 °C within the well plate
- Heating rate: from 37 °C (98.6 °F) up to 50 °C (122 °F) +/- 2 °C in 7 minutes
- Cooling rate: from 50 °C (122 °F) down to 40 °C (104 °F) +/- 2 °C in 16 minutes
- Existing adapters for loading: Abgene™ 96 Well 0.8 mL PP Deepwell Storage Plate