The technical data for the dilutor module is as follows:
Component | Technical data |
|---|---|
Dilutor valve | 6-port (6+) Ceramic (Alumina) valve for ¼” 28 fittings Maximum permissible pressure: 3 bar |
Dilutor syringe | 1 mL, scale 60 mm |
Transfer tubing | PFA tube ID 0.787 mm (1/32”) / OD 1.58 mm (1/16”) Including connection fittings and guiding wire |
Needle | Gauge 22, Pst 3, length 80 mm |
Wetted parts | Tubing from solution bottles to dilutor module:
Dilutor module syringe:
Transfer tubing from dilutor module to tool: PFA |