The technical data for the AccelerOme system is as follows:
Specification | Value |
|---|---|
Protection class (a) | Class I |
Over voltage category (b) | Category II |
Pollution degree (c) | 2 |
Moisture protection (d) | Normal (IPXO) |
Voltage | 100-240 VAC 50/60 Hz |
Fuse (PAL control board) | T6.3A/250V |
Fuse (Power inlet socket) | T4A/250V (2 pcs) |
a: Protection class I:
Protection class describes the insulating scheme used in the instrument to protect you from electrical shock. Class I identifies a single level of insulation between live parts (wires) and exposed conductive parts (metal panels), in which the exposed conductive parts are connected to a grounding system. This grounding system is connected to the third pin (ground pin) on the electrical power plug.
b: Over voltage category II:
Over voltage category II pertains to instruments that receive their electrical power from a local level such as an electrical wall outlet.
c: Pollution degree 2:
This is a measure of pollution in electrical circuits that might produce a reduction of the dielectric strength or surface resistivity.
Degree 2 normally refers only to non-conductive pollution.
Occasionally, however, a temporary conductivity caused by condensation is to be expected.
d: Moisture protection:
Normal (IPXO) – IPXO means that there is No Ingress Protection against any type of dripping or sprayed water. The X is a place holder to identify protection against dust, if applicable.