The emissions from the AccelerOme system are as follows:
Emission type | Emission value |
|---|---|
Sound pressure level (1 meter from the equipment in the direction of maximum sound pressure level) | <85 dBA |
The Accelerome instrument emits cooling air and waste water. The cooling air can contain hazardous substances. Therefore, it is important to observe the applicable regulations for health protection related to the substances processed in the AccelerOme instrument. The waste water requires correct disposal according to the national, regional, or local regulations for environmental protection. |