The AccelerOme instrument weighs about 85 kg (187.4 lb) when loaded and ready to use. Therefore, the load capacity of your table must be at least 200 kg (440.9 lb).
A place for the waste container is required below the table. This place must be below the level of the bottom of the instrument so that the liquid can drain away without backing up.
CAUTION
Heavy Object
If the instrument is placed on an uneven surface, it can fall. Not being careful while placing the instrument on the table can cause it to fall.
This can lead to a lower body injury to anyone in vicinity of the instrument.