The front door of the AccelerOme instrument is a part of the instrument's safety features. It is actively locked by means of an electromagnetic switch (solenoid). This lock mechanism prevents access to the instrument interior when the instrument is in use and protects you from interacting with the moving parts.