The thermal mixer module is used for orbital shaking and incubation of the samples. The AB0765 Abgene™ 96 Well 0.8mL Polypropylene Deepwell Storage Plate is used as the input sample plate. This module is used to shake and heat the batch-prepared samples over a long duration. The samples are shaken at a constant temperature.
Thermal mixer with attached sample plate
Thermal mixer with attached sample plate
To ensure good heat transfer between the heating plate of the thermal mixer and the sample plate, an adapter plate called the heat distributor is used upon which the sample plate is placed.
Attach sample plate to thermal mixer
Attach sample plate to thermal mixer
CAUTION
Hot Surface
Hot instrument parts can cause minor burns.
Do not touch the thermal mixer without the sample plate on top. Allow the adapter plate (heat distributor) of the thermal mixer to cool down before touching it.