There are two USB ports (USB 3.0) at the front of the instrument on the bottom right.
One can be used for the barcode scanner and one for the file transfer from the PC or USB drive.
There are two USB ports (USB 3.0) at the front of the instrument on the bottom right.
One can be used for the barcode scanner and one for the file transfer from the PC or USB drive.