There is an ethernet connection (RJ45) port and the socket for the power cord (100-240 VAC) on the back of the instrument on the bottom right.
The connector for the power cord has an integrated fuse.
| No. | Description | No. | Description |
|---|---|---|---|
1 | LAN cord for ethernet connection | 2 | Power cord |