Following are the minimum hardware and software configurations required for the AccelerOme Experiment Designer software 1.1 operation.
Computer
  • 3 GHz dual core processor with 8 GB RAM
  • NOTE
    To expedite the processing time for some workflows, Thermo Fisher Scientific recommends 16 GB of memory, even though the minimum requirement is 8 GB.
  • Resolution display: 1920 × 1080
  • NTFS file format supported
OS
Microsoft™ Windows™ 10
To install the Experiment Designer application, the user needs to log in to their Windows system as an administrator.