You can remove an instrument from the software, if needed.
Steps
  1. Click Instruments.
  2. Click Actions and choose Manage instruments.
  3. To remove an instrument, click the ellipsis (…) button.
  4. Click Remove.
  5. Click Remove to confirm.
  6. Click Done to close the dialog.