You can configure the Experiment Designer application to communicate with an AccelerOme instrument on the network to be able to send methods to connected instruments and view the status of the connected instruments. You can discover, add, or configure an instrument manually when the computer and the instrument are on a routed network and connected to the same subnet on the network.
NOTE
For discovery and communication to work, ensure that ports 8180, 9100, and UDP 15000 and 150001 are open.