Following are the parameters on the Assign Batches or Assign Sequences page of the Experiment Designer application.
Parameter | Description |
|---|---|
Actions | Select from: |
* View statistical power settings | * Displays information about the power settings. |
* Export as CSV | * Exports the information in CSV format. |
* Download report | * Downloads the report in PDF format. |
Color plate by: | Select from: |
*Sample | * Color the plate by sample information. |
* Study | * Color the plate by study information. |
* Factor Name | * Color the plate by factor information. |
* Batch | * Color the plate by batch information Only for TMT samples. |
* TMT Channel | * Color the plate by TMT channel information Only for TMT samples. |
Statistical Power Estimate | Select from: |
* Study Number | * View the statistical power estimate of the selected study. |
View Details | View the statistical power estimate details of the selected study. |
Process Order | The processing order of the sample. |
Well | The sample well or sample position on the plate. |
Batch | Batch number of the sample. Applies to TMT methods only. |
TMT Channel | TMT Channel number of the sample. Applies to TMT methods only. |
Study Name | The name of the study. |
Sample Name | The name of the sample. |
Factor | Factor value. |
On/Off | Randomize or unrandomize the samples. |
Next Step | Advance to the next page of information. |