Following are the parameters on the Edit Samples page of the Experiment Designer application.
Parameter | Description |
|---|---|
Actions | Select from: |
* Upload sample file | * Uploads a sample file in CSE or CSV format. |
* Download sample file | * Downloads a sample file. |
Method Name | Name of the sample method. |
Owner | (Optional) Name of the file creator or the last file editor. |
Kit Type | The available kits, listed by number of samples. |
Lot number | (Optional) The lot number. |
Samples | The number of samples in the experiment. |
Process Order | The processing order of the sample. |
Input Position | The input well position of the sample. |
Sample Name | The name of the sample. Update if necessary. |
Next Step | Advance to the next page of information. |