Following are the parameters for method file creation using the Experiment Designer application.
Parameter | Description |
|---|---|
Method Name | Name of the sample method. |
Creator | (Optional) Name of the file creator. |
Kit Details | The kit's type. |
Lot number | (Optional) The lot number. |
Samples | The number of samples in the experiment. |
Upload Sample File | (Optional) Upload sample information from one or more CSV/CSE format files. |
Create | Create a method file with the defined parameters. |
Cancel | Cancel method file creation. |