This screen on the Experiment Designer displays the history of experiments designed using the software, in a table format. The table entries display the method name, kit type, instrument information, and the date and time when the method file was saved or modified.
Selecting and right clicking a table entry provides the following options:
Parameter | Description |
|---|---|
Open file | Open a method file. |
Download method files | Download a method file. A shortcut to this action is also available as an icon in the rightmost column of the table. When a method file is saved, the saved file or folder opens and you are notified on the screen. |
View method report | View the method report. A shortcut to this action is also available as an icon in the rightmost column of the table. |
Download method report | Download a method report. A shortcut to this action is also available as an icon in the rightmost column of the table. When a method report is downloaded, the downloaded file or folder opens and you are notified on the screen. |
Delete file | Delete a method file. A shortcut to this action is also available as an icon in the rightmost column of the table. |
To download or delete multiple files, select the files by checking the check box to the left of the method name and click the Download or Delete buttons on the top right corner of the screen. To delete or download all files, check the check box in the table header of the check box column to select all files.