On the History screen of the Experiment Designer application you can view the method design history of the methods designed using the application and you can view a method file, download a method file, view a method report, download a method report, or delete a file. You can also download or delete multiple files at once.