Following are the parameters on the Settings page of the Experiment Designer application.
Parameter | Description |
|---|---|
Default Save Location | |
File Saved To | The folder path where the file is saved. |
Browse | Browse for the location to save the file. |
Save | Save the default file location. |
Cancel | Cancel action. |
Software Updates | |
Check for Update | Check for available software updates. |
Install Update | Install the latest available software version. |
Statistical Review | |
Minimum Biological Effect | Minimum biological effect value between 1 to 100% in 1% increments. |
Coefficient of Variation | Coefficient of variation value between 1 to 100% in 1% increments. |
Restore to System Default | Restore both assumption values to system default. |
Save | Save the edited assumption values. |
Cancel | Cancel action. |
File Name Convention | |
File Name Example | An example of a valid file name. |
Available Options | Options available to add to a file name. |
Add Custom Text | Custom created options to add to a file name. |
Restore to System Default | Restore file name convention to system default. |
Save | Save the updated file name convention. |
Cancel | Cancel action. |
About | |
Legal Notice | The legal notice about the Experiment Designer software. |
EULA | End User License Agreement for the Experiment Designer software. |