Use this setting to change the file name convention of a method file, if desired.
Steps
  1. Click Settings.
  2. Click File Name Convention.
  3. If there are text segments that you do not want to include in the file name, move them to the Available Options pane.
  4. If there are required text segments in Available Options, move them to the upper pane.
  5. If you require additional text segments, type them in the custom text field, and click the plus symbol.
  6. If necessary, move the segments to change the order.
  7. Click Save.
  8. (Optional) Click Restore to system default to restore the file name convention to system default.