If you do not upload the sample information to the Experiment Designer application while creating a method, you can enter it on the Edit Samples page.
Steps
- On the Edit Samples page, select each row and enter or edit the sample name, or select Actions > Upload Sample File.
- Click Next Step.
NOTE
All sample names in an experiment must be unique.
The number of samples in the uploaded file must match the number of samples in the experiment.
To save a sample file to your computer, after editing the sample names, select Actions > Download Sample File. You can upload this sample file to an experiment by selecting Actions > Upload Sample File from the Edit Samples page or by clicking Upload sample file in .csv or .txt format (optional) from the experiment type popup on the Experiment Designer home page.