You can design two types of methods using the Experiment Designer application:
- Label-Free Quantification (LFQ) method
- Tandem Mass Tag (TMT) method
Steps
- On the Experiment Designer home screen, click the experiment type, Label-Free or Tandem Mass Tag.
- To create an experiment based on an imported method file, see Import method.
- (Optional) Download the template files in CSV format (in a zip file) for Label-Free and Tandem Mass Tag experiment types by clicking on the three horizontal dots on the top left corner of the Label-Free or Tandem Mass Tag sample preparation experiment types.
- Enter the method name.
- (Optional) For Creator, type your name.
- For Kit Details, select the appropriate kit.
- (Optional) Type the lot number.
- For Samples:
- For Label-Free sample preparation, select the number of samples.
- For Tandem Mass Tag sample preparation, select the number of samples.