When you define your replicate groups (the study factor-value replicates), a randomization algorithm can be used to ensure the replicates are distributed among different batches and channels for TMT samples or the replicates are distributed in a random sequence order for LFQ samples.
By default, the order of samples is not randomized. The samples are filled in a top-left to bottom-right manner based on the order of the sample names.