The needle wash solutions are not included with the instrument. Prepare one liter of each of the following solutions:
Solution name | Solution composition |
|---|---|
Needle Wash Solution W1 | Needle Wash Solution W1 – 0.1% formic acid in water, LC-MS grade (for example, Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade) |
Needle Wash Solution W2 | Needle Wash Solution W2 – 50% methanol and 0.05% formic acid in water, LC-MS grade (for example, 1:1 v/v mix of needle wash solution W1 and Optima™ LC/MS Grade methanol) |