Prepare the AccelerOme system consumables and samples, as follows:
- The AccelerOme instrument needle wash solutions.
- One or more AccelerOme reagent kits (refer to the AccelerOme reagent kit guides).
- Prepare the biological samples according to the instructions in the AccelerOme reagent kit guides.
- For example, prepare 8 treatment-condition eukaryotic cell culture replicates that have more than 10^6 cells/mL each and 8 control-condition replicates of the same cell density.