Ensure that your laboratory table and the laboratory space meet the following requirements:
  • Have approximately 1 square meter or 10 square feet of space on the laboratory table that has 100 centimeters or approximately 3.3 feet of unobstructed vertical clearance.
  • The laboratory table must be horizontally-level, stable, resonance-free and away from heat and strong sunlight.
  • Have no obstructions in the front floor space of the table or on the left side of the table for instrument door access.
  • Have a minimum of 15 centimeters or 6 inches omnidirectional clearance to allow for air circulation.
  • Have a minimum door and hallway width of 80 centimeters or 31.5 inches to transport the packaged instrument through doors and hallways.
  • The laboratory table must have a load capacity of at least 200 kg or 441 lbs.
  • Have an electrical outlet (circuit capable of 20 A, 120 V or 10 A, 250V).
  • (Optional) Have an active ethernet port.
For full site requirements, refer to the laboratory requirements in the AccelerOme Preinstallation Guide.