You can import an existing method file created using the Experiment Designer application to the instrument.
Steps
- Click Import method file.
- To select the method file location, click Browse.
- Locate and select the method file in CSE or CSV format from your computer.
- Click Upload.
If a method file has an incorrect format, an error message is displayed upon uploading the file.
The Download Template Files option in the Label-Free and Tandem Mass Tag sample preparation types allows you to download the available file templates. The See Help option directs you to the Experiment Designer Help.