Experiment Designer is the desktop software application used to design method files for an experiment.
Steps
- Insert the product USB drive into your computer.
- Start the installer by running the Experiment Designer msi installer file from the USB drive.
- Follow the installation wizard.
NOTE
The .NET framework version needs to be 4.7.2 or later for successful installation.