AccelerOme™ is an Automated Sample Preparation Platform (ASPP) that includes the AccelerOme instrument and the reagents that simplify and standardize mass spectrometer sample preparation.
The system processes samples for both label-free and tandem mass tag (TMT) based quantitative proteomics in approximately 4 to 6 hours.
The system includes the Experiment Designer software application for designing the experiment method files. This application supports label-free and TMT chemistry for sample types.
This document describes how to set up a new AccelerOme instrument, design an Experiment Designer experiment method file, and run that method file on the AccelerOme instrument.
This Quick Start Guide contains the following topics:
Install consumables and system components
Load the needle wash solutions
Set up the instrument interface
Install the Experiment Designer software
Run an experiment using a method file