Provide a waste container with a minimum volume of 4 liters with a duct cover that fits the waste line tightly.
Provide a secondary container to hold the main waste container to prevent waste fluid overflow.