The instrument dimensions and mass are stated as follows:
Depth (Y-axis) | Width (X-axis) | Height (Z-axis) | Mass | ||||
|---|---|---|---|---|---|---|---|
cm | in | cm | in | cm | in | kg | lbs |
75.6 | 30 | 63.5 | 25 | 88.5 | 35 | 75 | 165 |
NOTE
Cable dimensions are not included.
The mass of the instrument is with packaging and without reagents. Fully loaded instrument is approximately 85 kg or 187 lbs.
Ensure you meet the following space and load requirements:
- Have a space of at least 9 cm (3.5 in) in front of the instrument as the instrument door opens to the front.
- Have at least 15 cm (6 in) of clearance behind the instrument.
- Have at least 92 cm (3 ft) of clearance above the instrument.
- Ensure the laboratory table can support the AccelerOme instrument. Remember that additional components loaded within the instrument add to the total weight.
- Ensure that your work area is free of vibration from the nearby equipments.
- Ensure you have sufficient ventilation in your work area.
- Ensure a minimum floor space of 120 x 105 cm with a height of at least 100 cm.
- Ensure a minimum distance of 40 cm to the wall on the left side of the instrument so that the door to the solution compartment can be opened completely.
- Ensure a minimum distance of approximately 15 cm to the right, back and top of the instrument to guarantee sufficient air circulation for the exhaust air and to avoid any heat buildup.