Thermo Fisher Scientific systems operate safely and reliably under specific environmental conditions. If you use the system in a manner not specified by the manufacturer, the protective features of the equipment might get impaired. If you do not maintain the system according to the specifications listed in this guide, failures of many types, including personal injury or death, might occur. The repair of instrument failures caused by improper use such as not specified by the manufacturer is specifically excluded from the standard warranty and service contract coverage.
Power rating
AccelerOme:
100-240 V AC +/-10%; 50/60 Hz; 250 VA
Detailed instrument specifications are available in the AccelerOme User Guide.